Skip to Content
MilliporeSigma
2-(Diphenylphosphino)ethylamine ≥95.0% (GC)

2-(Diphenylphosphino)ethylamine

2-(Diphenylphosphino)ethylamine ≥95.0% (GC)
Synonym(s):
(2-Aminoethyl)diphenylphosphine, 1-Amino-2-(diphenylphosphino)ethane
Empirical Formula (Hill Notation):
C14H16NP
CAS Number:
Molecular Weight:
229.26

Products

Brand
Formula Weight
Physical Form
Purity
Functional Group
Markush Class
Markush Group
Reaction Type
Reagent Type
Showing 1-1 of 1
Compare
Product Number
Description
Pricing