Skip to Content
MilliporeSigma
2-(Dimethylamino)ethanethiol hydrochloride 95%

2-(Dimethylamino)ethanethiol hydrochloride

2-(Dimethylamino)ethanethiol hydrochloride 95%
Synonym(s):
MEDA
Linear Formula:
(CH3)2NCH2CH2SH · HCl
CAS Number:
Molecular Weight:
141.66
EC Number:

Products

Product Category
Formula Weight
Melting Point (°C)
Physical Form
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing