Skip to Content
MilliporeSigma
2′-Deoxyadenosine monohydrate powder, BioReagent, suitable for cell culture

2′-Deoxyadenosine monohydrate

2′-Deoxyadenosine monohydrate powder, BioReagent, suitable for cell culture
Synonym(s):
9-(2-Deoxy-β-D-ribofuranosyl)adenine, Adenine deoxyriboside
Empirical Formula (Hill Notation):
C10H13N5O3 · H2O
CAS Number:
Molecular Weight:
269.26
EC Number:

Products

Brand
Biological Source
Formula Weight
Physical Form
Product Line
Purity
Technique
Showing 1-2 of 2