Skip to Content
MilliporeSigma
2-cis,4-trans-Abscisic acid synthetic, 98%

2-cis,4-trans-Abscisic acid

2-cis,4-trans-Abscisic acid synthetic, 98%
Synonym(s):
(±)-Abscisic acid, (2Z,4E)-5-(1-Hydroxy-2,6,6-trimethyl-4-oxo-2-cyclohexen-1-yl)-3-methyl-2,4-pentadienoic acid, ABA, Dormin
Empirical Formula (Hill Notation):
C15H20O4
CAS Number:
Molecular Weight:
264.32

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing