![2-(Chloromethyl)allyl-trimethylsilane 97%](/deepweb/assets/sigmaaldrich/product/structures/614/476/d79c119e-0e2c-4837-959e-cef5fe8d3a9c/640/d79c119e-0e2c-4837-959e-cef5fe8d3a9c.png)
2-(Chloromethyl)allyl-trimethylsilane
![2-(Chloromethyl)allyl-trimethylsilane 97%](/deepweb/assets/sigmaaldrich/product/structures/614/476/d79c119e-0e2c-4837-959e-cef5fe8d3a9c/640/d79c119e-0e2c-4837-959e-cef5fe8d3a9c.png)
Synonym(s):
2-(Chloromethyl)-3-trimethylsilyl-1-propene, 2-(Trimethylsilylmethyl)allyl chloride
Linear Formula:
(CH3)3SiCH2C(CH2Cl)=CH2
CAS Number:
Molecular Weight:
162.73
Products
Product Category
Brand
Boiling Point (°C)
Formula Weight
Physical Form
Purity
Quality Segment
Functional Group
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing