Skip to Content
MilliporeSigma
2′-Chloroacetophenone 97%

2′-Chloroacetophenone

2′-Chloroacetophenone 97%
Synonym(s):
2′-Chloroacetophenone
Linear Formula:
ClC6H4COCH3
CAS Number:
Molecular Weight:
154.59

Products

Product Category
Brand
Boiling Point (°C)
Formula Weight
Physical Form
Purity
Quality Segment
Markush Class
Markush Group
Showing 1-2 of 2
Compare
Product Number
Description
Pricing