Skip to Content
MilliporeSigma
2-Chloro-3-methylquinoxaline 97%

2-Chloro-3-methylquinoxaline

2-Chloro-3-methylquinoxaline 97%
Empirical Formula (Hill Notation):
C9H7ClN2
CAS Number:
Molecular Weight:
178.62

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Functional Group
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing