Skip to Content
MilliporeSigma
2-Chloro-1,3-dimethylimidazolidinium hexafluorophosphate 98%

2-Chloro-1,3-dimethylimidazolidinium hexafluorophosphate

2-Chloro-1,3-dimethylimidazolidinium hexafluorophosphate 98%
Synonym(s):
2-Chloro-4,5-dihydro-1,3-dimethyl-1H-imidazolium hexafluorophosphate, CIP
Empirical Formula (Hill Notation):
C5H10ClF6N2P
CAS Number:
Molecular Weight:
278.56

Products

Brand
Formula Weight
Melting Point (°C)
Purity
Functional Group
Application
Reaction Type
Showing 1-1 of 1
Compare
Product Number
Description
Pricing