Skip to Content
MilliporeSigma
vibrant-m

(2-Butylbenzofuran-3-yl)(4-hydroxy-3,5-diiodophenyl)methanone

vibrant-m
Synonym(s):
NSC 85437
Empirical Formula (Hill Notation):
C19H16I2O3
CAS Number:
Molecular Weight:
546.14

Products

Formula Weight
Manufacturer
Physical Form
Format
Application
API Family
Reference Material Type
Special Grade