![2-Bromoquinoline 95%](/deepweb/assets/sigmaaldrich/product/structures/270/939/c1509a54-5276-4145-8e89-c0e21814d9e3/640/c1509a54-5276-4145-8e89-c0e21814d9e3.png)
2-Bromoquinoline
![2-Bromoquinoline 95%](/deepweb/assets/sigmaaldrich/product/structures/270/939/c1509a54-5276-4145-8e89-c0e21814d9e3/640/c1509a54-5276-4145-8e89-c0e21814d9e3.png)
Empirical Formula (Hill Notation):
C9H6BrN
CAS Number:
Molecular Weight:
208.05
Products
Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Functional Group
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing