Skip to Content
MilliporeSigma
2-Bromopyridine-4-carboxylic acid 97%

2-Bromopyridine-4-carboxylic acid

2-Bromopyridine-4-carboxylic acid 97%
Synonym(s):
2-Bromoisonicotinic acid
Empirical Formula (Hill Notation):
C6H4BrNO2
CAS Number:
Molecular Weight:
202.01

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Functional Group
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing