Skip to Content
MilliporeSigma
2-(Bromomethyl)benzonitrile ≥97%

2-(Bromomethyl)benzonitrile

2-(Bromomethyl)benzonitrile ≥97%
Synonym(s):
α-Bromo-o-tolunitrile, 2-Cyanobenzyl bromide
Linear Formula:
BrCH2C6H4CN
CAS Number:
Molecular Weight:
196.04

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Functional Group
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing