Skip to Content
MilliporeSigma
2-Bromohexadecanoic acid ~97%

2-Bromohexadecanoic acid

2-Bromohexadecanoic acid ~97%
Synonym(s):
2-Bromopalmitic acid
Linear Formula:
CH3(CH2)13CH(Br)CO2H
CAS Number:
Molecular Weight:
335.32
EC Number:

Products

Product Category
Formula Weight
Gene Alias
Melting Point (°C)
Physical Form
Purity
Functional Group
Markush Class
Markush Group
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing