Skip to Content
MilliporeSigma
2-(Boc-amino)ethanethiol 97%

2-(Boc-amino)ethanethiol

2-(Boc-amino)ethanethiol 97%
Synonym(s):
tert-Butyl N-(2-mercaptoethyl)carbamate
Linear Formula:
HSCH2CH2NHCO2C(CH3)3
CAS Number:
Molecular Weight:
177.26

Products

Product Category
Boiling Point (°C)
Formula Weight
Purity
Functional Group
Markush Class
Markush Group
Reagent Type
Showing 1-1 of 1
Compare
Product Number
Description
Pricing