Skip to Content
MilliporeSigma
vibrant-m

2-(Biphenyl-4-yl)propionic acid

vibrant-m
Synonym(s):
Biprofen, 2-(4-Biphenylyl)propionic acid, NSC 16300
Empirical Formula (Hill Notation):
C15H14O2
CAS Number:
Molecular Weight:
226.27

Products

Formula Weight
Manufacturer
Physical Form
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing