Skip to Content
MilliporeSigma
2-Benzyloxy-1,3-propanediol 99%

2-Benzyloxy-1,3-propanediol

2-Benzyloxy-1,3-propanediol 99%
Synonym(s):
2-O-Benzylglycerol, Glycerol 2-benzyl ether
Linear Formula:
C6H5CH2OCH(CH2OH)2
CAS Number:
Molecular Weight:
182.22

Products

Brand
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Purity
Markush Class
Markush Group
Monomer Type
Showing 1-1 of 1
Compare
Product Number
Description
Pricing