Skip to Content
MilliporeSigma
2-Azetidinone 98%

2-Azetidinone

2-Azetidinone 98%
Synonym(s):
β-Propiolactam, 2-Azacyclobutanone
Empirical Formula (Hill Notation):
C3H5NO
CAS Number:
Molecular Weight:
71.08

Products

Product Category
Brand
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing