Skip to Content
MilliporeSigma
2-Aminoterephthalic acid 99%

2-Aminoterephthalic acid

2-Aminoterephthalic acid 99%
Linear Formula:
H2NC6H3-1,4-(CO2H)2
CAS Number:
Molecular Weight:
181.15

Products

Product Category
Formula Weight
Melting Point (°C)
Physical Form
Purity
Application
Markush Class
Markush Group
Amino Acid
Amino Acid Type
Reaction Type
Showing 1-1 of 1
Compare
Product Number
Description
Pricing