Skip to Content
MilliporeSigma
2-Aminopyrimidine-5-carboxaldehyde 97%

2-Aminopyrimidine-5-carboxaldehyde

2-Aminopyrimidine-5-carboxaldehyde 97%
Linear Formula:
C4H2N2NH2CHO
CAS Number:
Molecular Weight:
123.11

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Showing 1-1 of 1
Compare
Product Number
Description
Pricing