![2-Aminoheptane 99%](/deepweb/assets/sigmaaldrich/product/structures/131/635/96c41194-61d9-4196-8bf5-531f42463fe3/640/96c41194-61d9-4196-8bf5-531f42463fe3.png)
2-Aminoheptane
![2-Aminoheptane 99%](/deepweb/assets/sigmaaldrich/product/structures/131/635/96c41194-61d9-4196-8bf5-531f42463fe3/640/96c41194-61d9-4196-8bf5-531f42463fe3.png)
Synonym(s):
1-Methylhexylamine, 2-Heptylamine, 2-Aminoheptane
Linear Formula:
CH3(CH2)4CH(NH2)CH3
CAS Number:
Molecular Weight:
115.22
Products
Product Category
Brand
Boiling Point (°C)
Formula Weight
Physical Form
Purity
Quality Segment
Markush Class
Markush Group
Showing 1-2 of 2
Compare
Product Number
Description
Pricing