Skip to Content
MilliporeSigma
2-Aminobenzyl alcohol certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

2-Aminobenzyl alcohol

2-Aminobenzyl alcohol certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Synonym(s):
2-(Hydroxymethyl)aniline
Linear Formula:
H2NC6H4CH2OH
CAS Number:
Molecular Weight:
123.15

Products

Product Category
Brand
Boiling Point (°C)
Formula Weight
Manufacturer
Melting Point (°C)
Physical Form
Product Line
Purity
Quality Segment
Application
Markush Class
Markush Group
Special Grade