Skip to Content
MilliporeSigma
2-Amino-4-bromophenol ≥94%

2-Amino-4-bromophenol

2-Amino-4-bromophenol ≥94%
Synonym(s):
4-Bromo-2-aminophenol, NSC 523846
Empirical Formula (Hill Notation):
C6H6BrNO
CAS Number:
Molecular Weight:
188.02

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Functional Group
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing