Skip to Content
MilliporeSigma
2-Acrylamido-2-methyl-1-propanesulfonic acid 99%

2-Acrylamido-2-methyl-1-propanesulfonic acid

2-Acrylamido-2-methyl-1-propanesulfonic acid 99%
Linear Formula:
H2C=CHCONHC(CH3)2CH2SO3H
CAS Number:
Molecular Weight:
207.25
EC Number:

Products

Product Category
Formula Weight
Melting Point (°C)
Purity
Markush Class
Markush Group
Monomer Type
Showing 1-1 of 1
Compare
Product Number
Description
Pricing