Skip to Content
MilliporeSigma
2,6-Dimethylaniline 99%

2,6-Dimethylaniline

2,6-Dimethylaniline 99%
Synonym(s):
2,6-Xylidine, 2-Amino-1,3-dimethylbenzene, 2-Amino-m-xylene, 2,6-Dimethylaniline
Linear Formula:
2,6-(CH3)2C6H3NH2
CAS Number:
Molecular Weight:
121.18

Products

Product Category
Brand
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Physical Form
Product Line
Purity
Format
Quality Segment
Application
Markush Class
Markush Group
Package Size
Reference Material Type
Special Grade
Technique
Showing 1-4 of 4