Skip to Content
MilliporeSigma
2,6-Difluorobenzyl bromide 97%

2,6-Difluorobenzyl bromide

2,6-Difluorobenzyl bromide 97%
Synonym(s):
α-Bromo-2,6-difluorotoluene
Linear Formula:
F2C6H3CH2Br
CAS Number:
Molecular Weight:
207.02
EC Number:

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Functional Group
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing