Skip to Content
MilliporeSigma
2,6-Dichloroindophenol sodium salt hydrate suitable for vitamin C determination, BioReagent

2,6-Dichloroindophenol sodium salt hydrate

2,6-Dichloroindophenol sodium salt hydrate suitable for vitamin C determination, BioReagent
Synonym(s):
2,6-Dichloro-N-(4-hydroxyphenyl)-1,4-benzoquinoneimine sodium salt, 2,6-Dichlorobenzenone-indophenol sodium salt, 2,6-Dichlorophenolindophenol sodium salt hydrate, DCIP, DPIP
Empirical Formula (Hill Notation):
C12H6Cl2NNaO2 · xH2O
CAS Number:
Molecular Weight:
290.08 (anhydrous basis)
EC Number:

Products

Product Category
Formula Weight
Physical Form
Product Line
Application
Showing 1-1 of 1
Compare
Product Number
Description
Pricing