Skip to Content
MilliporeSigma
2-(6-Bromohexyloxy)tetrahydro-2H-pyran 97%

2-(6-Bromohexyloxy)tetrahydro-2H-pyran

2-(6-Bromohexyloxy)tetrahydro-2H-pyran 97%
Synonym(s):
1-Bromo-6-tetrahydropyranyloxyhexane, 6-Tetrahydropyraryloxy-1-bromohexane
Empirical Formula (Hill Notation):
C11H21BrO2
CAS Number:
Molecular Weight:
265.19
EC Number:

Products

Product Category
Brand
Boiling Point (°C)
Formula Weight
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing