Skip to Content
MilliporeSigma
2,6-Bis(chloromethyl)pyridine 99%

2,6-Bis(chloromethyl)pyridine

2,6-Bis(chloromethyl)pyridine 99%
Synonym(s):
α,α′-Dichloro-2,6-lutidine
Empirical Formula (Hill Notation):
C7H7Cl2N
CAS Number:
Molecular Weight:
176.04

Products

Product Category
Formula Weight
Melting Point (°C)
Physical Form
Purity
Functional Group
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing