Skip to Content
MilliporeSigma
2,6-Bis(bromomethyl)naphthalene 95%

2,6-Bis(bromomethyl)naphthalene

2,6-Bis(bromomethyl)naphthalene 95%
Empirical Formula (Hill Notation):
C12H10Br2
CAS Number:
Molecular Weight:
314.02

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Functional Group
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing