Skip to Content
MilliporeSigma
25-Hydroxyvitamin D3 (6,19,19-d3) solution 5 μg/mL in ethanol, 97 atom % D, 98% (CP)

25-Hydroxyvitamin D3 (6,19,19-d3) solution

25-Hydroxyvitamin D3 (6,19,19-d3) solution 5 μg/mL in ethanol, 97 atom % D, 98% (CP)
Synonym(s):
25-Hydroxycholecalciferol (6,19,19-d3) solution, Deuterated 25-hydroxyvitamin D3
Empirical Formula (Hill Notation):
C27D3H41O2
CAS Number:
Molecular Weight:
403.66
EC Number:

Products

Product Category
Brand
Color
Formula Weight
Physical Form
Purity
Isotope
Application
Markush Class
Markush Group
Technique