Skip to Content
MilliporeSigma
2,5-Di-tert-butylhydroquinone 99%

2,5-Di-tert-butylhydroquinone

2,5-Di-tert-butylhydroquinone 99%
Synonym(s):
2,5-Di-tert-butylhydroquinone
Linear Formula:
2,5-[CH3)3C]2C6H2-1,4-(OH)2
CAS Number:
Molecular Weight:
222.32

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Markush Class
Markush Group
Showing 1-2 of 2
Compare
Product Number
Description
Pricing