Skip to Content
MilliporeSigma
2,5-Di-tert-butylaniline 99%

2,5-Di-tert-butylaniline

2,5-Di-tert-butylaniline 99%
Linear Formula:
[(CH3)3C]2C6H3NH2
CAS Number:
Molecular Weight:
205.34

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing