Skip to Content
MilliporeSigma
2,5-Dimethyl-celecoxib ≥98% (HPLC)

2,5-Dimethyl-celecoxib

2,5-Dimethyl-celecoxib ≥98% (HPLC)
Synonym(s):
2,5-Dimethylcelecoxib, 4-[5-(2,5-dimethylphenyl)-3-(trifluoromethyl)-1H-pyrazol-1-yl]-benzenesulfonamide
Empirical Formula (Hill Notation):
C18H16F3N3O2S
CAS Number:
Molecular Weight:
395.40

Products

Product Category
Brand
Color
Formula Weight
Physical Form
Purity
Research Area
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing