Skip to Content
MilliporeSigma
2,4-Difluoro-α-(1H-1,2,4-triazolyl)acetophenone 95%

2,4-Difluoro-α-(1H-1,2,4-triazolyl)acetophenone

2,4-Difluoro-α-(1H-1,2,4-triazolyl)acetophenone 95%
Synonym(s):
1-(2,4-Difluorophenyl)-2-(1H-1,2,4-triazol-1-yl)ethanone
Empirical Formula (Hill Notation):
C10H7F2N3O
CAS Number:
Molecular Weight:
223.18

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Functional Group
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing