Skip to Content
MilliporeSigma
2,4-Dichlorophenylboronic acid ≥95%

2,4-Dichlorophenylboronic acid

2,4-Dichlorophenylboronic acid ≥95%
Synonym(s):
2,4-Dichlorobenzeneboronic acid
Linear Formula:
Cl2C6H3B(OH)2
CAS Number:
Molecular Weight:
190.82

Products

Brand
Formula Weight
Melting Point (°C)
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing