![2,4-Dichlorobenzyl alcohol impurity C European Pharmacopoeia (EP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/166/778/8a42b992-c947-43ac-b35f-5703e4d7d463/640/8a42b992-c947-43ac-b35f-5703e4d7d463.png)
2,4-Dichlorobenzyl alcohol impurity C
![2,4-Dichlorobenzyl alcohol impurity C European Pharmacopoeia (EP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/166/778/8a42b992-c947-43ac-b35f-5703e4d7d463/640/8a42b992-c947-43ac-b35f-5703e4d7d463.png)
Synonym(s):
3,4-Dichlorobenzyl alcohol
Linear Formula:
Cl2C6H3CH2OH
CAS Number:
Molecular Weight:
177.03
Products
Boiling Point (°C)
Formula Weight
Manufacturer
Melting Point (°C)
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing