![2,4-Dichlorobenzaldehyde 99%](/deepweb/assets/sigmaaldrich/product/structures/335/760/5849475b-eccd-44a2-90a1-4a89cf69fa87/640/5849475b-eccd-44a2-90a1-4a89cf69fa87.png)
2,4-Dichlorobenzaldehyde
![2,4-Dichlorobenzaldehyde 99%](/deepweb/assets/sigmaaldrich/product/structures/335/760/5849475b-eccd-44a2-90a1-4a89cf69fa87/640/5849475b-eccd-44a2-90a1-4a89cf69fa87.png)
Synonym(s):
2,4-Dichlorobenzaldehyde
Linear Formula:
2,4-(Cl)2C6H3CHO
CAS Number:
Molecular Weight:
175.01
Products
Product Category
Brand
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Functional Group
Markush Class
Markush Group
Showing 1-2 of 2
Compare
Product Number
Description
Pricing