Skip to Content
MilliporeSigma
2,4-Diamino-6,7-diisopropylpteridine phosphate salt ≥98%

2,4-Diamino-6,7-diisopropylpteridine phosphate salt

2,4-Diamino-6,7-diisopropylpteridine phosphate salt ≥98%
Empirical Formula (Hill Notation):
C12H18N6 · H3PO4
CAS Number:
Molecular Weight:
344.31
EC Number:

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing