Skip to Content
MilliporeSigma
2-[[(4-Chloro-3,5-dimethyl-2-pyridinyl)methyl]thio]-6-methoxy-1H-benzimidazole certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

2-[[(4-Chloro-3,5-dimethyl-2-pyridinyl)methyl]thio]-6-methoxy-1H-benzimidazole

2-[[(4-Chloro-3,5-dimethyl-2-pyridinyl)methyl]thio]-6-methoxy-1H-benzimidazole certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Synonym(s):
5-Methoxy-2-[[(4-chloro-3,5-dimethylpyridin-2-yl)methyl]sulfanyl]-1H-benzimidazole
Empirical Formula (Hill Notation):
C16H16ClN3OS
CAS Number:
Molecular Weight:
333.84

Products

Brand
Formula Weight
Manufacturer
Product Line
Quality Segment
Application
Special Grade