Skip to Content
MilliporeSigma
2,4,6-Triphenylpyrylium tetrafluoroborate 98%

2,4,6-Triphenylpyrylium tetrafluoroborate

2,4,6-Triphenylpyrylium tetrafluoroborate 98%
Synonym(s):
2,4,6-Triphenylpyrylium tetrafluoroborate
Empirical Formula (Hill Notation):
C23H17BF4O
CAS Number:
Molecular Weight:
396.19

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Markush Class
Markush Group
Reaction Type
Reagent Type
Showing 1-2 of 2