Skip to Content
MilliporeSigma
2′,4′,6′-Trimethoxyacetophenone 97%

2′,4′,6′-Trimethoxyacetophenone

2′,4′,6′-Trimethoxyacetophenone 97%
Empirical Formula (Hill Notation):
C11H14O4
CAS Number:
Molecular Weight:
210.23

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing