Skip to Content
MilliporeSigma
2,4,6-Triisopropylbenzenesulfonyl hydrazide 90%

2,4,6-Triisopropylbenzenesulfonyl hydrazide

2,4,6-Triisopropylbenzenesulfonyl hydrazide 90%
Synonym(s):
2,4,6-Triisopropylbenzenesulfonohydrazide, TPSH, Trisylhydrazide
Linear Formula:
[(CH3)2CH]3C6H2SO2NHNH2
CAS Number:
Molecular Weight:
298.44
EC Number:

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing