Skip to Content
MilliporeSigma
2,4,6-Tribromoanisole 99%

2,4,6-Tribromoanisole

2,4,6-Tribromoanisole 99%
Synonym(s):
2,4,6-Tribromo-1-methoxybenzene
Linear Formula:
Br3C6H2OCH3
CAS Number:
Molecular Weight:
344.83

Products

Product Category
Brand
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Physical Form
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing