Skip to Content
MilliporeSigma
2-[4-(1-Hydroxy-2-methylpropyl)phenyl]propanoic acid pharmaceutical impurity standard

2-[4-(1-Hydroxy-2-methylpropyl)phenyl]propanoic acid

2-[4-(1-Hydroxy-2-methylpropyl)phenyl]propanoic acid pharmaceutical impurity standard
Synonym(s):
1-Hydroxyibuprofen, 2-[4-(1-Hydroxy-2-methylpropyl)phenyl]propionic acid
Empirical Formula (Hill Notation):
C13H18O3
CAS Number:
Molecular Weight:
222.28

Products

Formula Weight
Purity
Format
Quality Segment
Application
Showing 1-1 of 1