Skip to Content
MilliporeSigma
23-Hydroxybetulinic acid ≥98% (HPLC)

23-Hydroxybetulinic acid

23-Hydroxybetulinic acid ≥98% (HPLC)
Synonym(s):
(3β,4α)-3,23-Dihydroxylup-20(29)-en-28-oic acid, 3β,23-Dihydroxylup-20(29)-en-28-oic acid, 3aH-Cyclopenta[a]chrysene, lup-20(29)-en-28-oic acid deriv., Anemosapogenin
Empirical Formula (Hill Notation):
C30H48O4
CAS Number:
Molecular Weight:
472.70

Products

Product Category
Brand
Color
Formula Weight
Physical Form
Purity
Research Area
Showing 1-1 of 1
Compare
Product Number
Description
Pricing