Skip to Content
MilliporeSigma
2,3-Diethyl-5-methylpyrazine 99%

2,3-Diethyl-5-methylpyrazine

2,3-Diethyl-5-methylpyrazine 99%
Empirical Formula (Hill Notation):
C9H14N2
CAS Number:
Molecular Weight:
150.22
EC Number:

Products

Product Category
Biological Source
Formula Weight
Purity
Agency
Quality Segment
Application
Markush Class
Markush Group
Organoleptic Class
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing