Skip to Content
MilliporeSigma
2,3,5-Trimethylpyrazine natural, ≥95%, FG

2,3,5-Trimethylpyrazine

2,3,5-Trimethylpyrazine natural, ≥95%, FG
Empirical Formula (Hill Notation):
C7H10N2
CAS Number:
Molecular Weight:
122.17
EC Number:

Products

Product Category
Sustainability
Biological Source
Boiling Point (°C)
Feature
Formula Weight
Physical Form
Purity
Agency
Quality Segment
Application
Markush Class
Markush Group
Organoleptic Class
Special Grade
Showing 1-3 of 3
Compare
Product Number
Description
Pricing