Skip to Content
MilliporeSigma
2,3,4′,5-Tetrahydroxystilbene 2-glucoside phyproof® Reference Substance

2,3,4′,5-Tetrahydroxystilbene 2-glucoside

2,3,4′,5-Tetrahydroxystilbene 2-glucoside phyproof® Reference Substance
Synonym(s):
2,3,5,4′-Tetrahydroxystilbene 2-O-β-D-glucoside, 2,4-Dihydroxy-6-[(1E)-2-(4-hydroxyphenyl)ethenyl]phenyl β-D-Glucopyranoside, EH-201, THSG, Tetrahydroxystilbene glucoside
Empirical Formula (Hill Notation):
C20H22O9
CAS Number:
Molecular Weight:
406.38

Products

Formula Weight
Manufacturer
Physical Form
Product Line
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing