Skip to Content
MilliporeSigma
2,3,4,5-Tetrafluorobenzoyl chloride 98%

2,3,4,5-Tetrafluorobenzoyl chloride

2,3,4,5-Tetrafluorobenzoyl chloride 98%
Linear Formula:
HC6F4COCl
CAS Number:
Molecular Weight:
212.53

Products

Product Category
Brand
Formula Weight
Physical Form
Purity
Functional Group
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing