Skip to Content
MilliporeSigma
2-(2-Hydroxyphenyl)-1H-benzimidazole 95%

2-(2-Hydroxyphenyl)-1H-benzimidazole

2-(2-Hydroxyphenyl)-1H-benzimidazole 95%
Synonym(s):
2-(1H-Benzimidazole-2-yl)phenol, 2-(1H-Benzimidazole-Z-yl)phenol, 2-(1H-Benzoimidazol-2-yl)-phenol
Empirical Formula (Hill Notation):
C13H10N2O
CAS Number:
Molecular Weight:
210.23

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing